Assay Details
Target Gene Details
Entrez Gene ID: | 67046 |
Gene Name: | TBC1 domain family, member 7 |
Gene Aliases: |
2610009C09Rik |
Location: |
Chr.13:43151740-43171501 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d7 | NM_001252639.1 | 8 | 1105 | NP_001239568.1 |
| NM_001252640.1 | 8 | 991 | NP_001239569.1 | |
| NM_025935.3 | 8 | 1085 | NP_080211.1 | |
| AK003271.1 | 4 | 483 | ||
| AK011356.1 | 8 | 944 | BAB27564.1 | |
| AK156543.1 | 8 | 1013 | BAE33751.1 | |
| BC125307.1 | 8 | 932 | ||
| BC125309.1 | 8 | 932 | ||
| BC145156.1 | 8 | 838 | ||
| BC145157.1 | 8 | 952 | ||
| DN174647.1 | 5 | 520 |
More Information
Set Membership: |
|

Genomic Map