Assay Details
Target Gene Details
Entrez Gene ID: | 19766 |
Gene Name: | receptor (TNFRSF)-interacting serine-threonine kinase 1 |
Gene Aliases: |
D330015H01Rik, RIP, Rinp, Rip1 |
Location: |
Chr.13:34000844-34035170 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ripk1 | NM_009068.3 | 1 | 39 | NP_033094.3 |
| AK030959.1 | BAC27194.1 | |||
| AK048989.1 | ||||
| AK156803.1 | BAE33860.1 | |||
| BC050905.1 | 1 | 39 | AAH50905.2 | |
| BC054542.1 | 1 | 39 | AAH54542.1 | |
| BC058162.1 | AAH58162.1 |
More Information
Set Membership: |
|

Genomic Map