Assay Details
Target Gene Details
Entrez Gene ID: | 68346 |
Gene Name: | sirtuin 5 |
Gene Aliases: |
0610012J09Rik, 1500032M05Rik, AV001953 |
Location: |
Chr.13:43365439-43395203 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sirt5 | NM_178848.3 | NP_849179.1 | ||
| AK002609.1 | ||||
| AK005346.1 | ||||
| BC031770.1 | AAH31770.1 | |||
| BC087898.1 | AAH87898.1 |

Genomic Map