Assay Details
Target Gene Details
Entrez Gene ID: | 75607 |
Gene Name: | WNK lysine deficient protein kinase 2 |
Gene Aliases: |
1810073P09Rik, AW122246, ESTM15, X83337, mKIAA1760 |
Location: |
Chr.13:49036302-49148574 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wnk2 | NM_001290313.1 | 30 | 7061 | NP_001277242.1 |
| NM_029361.4 | 28 | 6791 | NP_083637.2 | |
| AI464179.1 | 1 | 142 | ||
| AK007980.1 | 5 | 689 | BAE43209.1 | |
| AK147550.1 | 28 | 6496 | BAE27991.1 | |
| BC046464.1 | 21 | 4716 | AAH46464.1 | |
| BC055795.1 | 9 | 1812 | AAH55795.1 | |
| BC060187.1 | 19 | 4092 | AAH60187.1 |
More Information
Set Membership: |
|

Genomic Map