Assay Details
Target Gene Details
Entrez Gene ID: | 19766 |
Gene Name: | receptor (TNFRSF)-interacting serine-threonine kinase 1 |
Gene Aliases: |
D330015H01Rik, RIP, Rinp, Rip1 |
Location: |
Chr.13:34000844-34035170 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ripk1 | NM_009068.3 | 5 | 751 | NP_033094.3 |
| AK030959.1 | 5 | 982 | BAC27194.1 | |
| AK156803.1 | 5 | 1016 | BAE33860.1 | |
| BC050905.1 | 5 | 751 | AAH50905.2 | |
| BC054542.1 | 5 | 751 | AAH54542.1 | |
| BC058162.1 | 5 | 937 | AAH58162.1 | |
| U25995.1 | 4 | 666 | AAB60487.1 |
More Information
Set Membership: |
|

Genomic Map