Assay Details
Target Gene Details
Entrez Gene ID: | 73710 |
Gene Name: | tubulin, beta 2B class IIB |
Gene Aliases: |
2410129E14Rik, brdp |
Location: |
Chr.13:34127008-34130354 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb2b | NM_023716.2 | NP_076205.1 | ||
| AK010786.1 | BAB27182.1 | |||
| AK168908.1 | BAE40722.1 | |||
| BC138935.1 | ||||
| BC138936.1 |
More Information
Set Membership: |
|

Genomic Map