Assay Details
Target Gene Details
Entrez Gene ID: | 73412 |
Gene Name: | NME/NM23 family member 8 |
Gene Aliases: |
1700056P15Rik, Sptrx-2, Txndc3 |
Location: |
Chr.13:19645061-19697760 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nme8 | NM_001167909.1 | 11 | 1089 | NP_001161381.1 |
| NM_181591.3 | 11 | 1089 | NP_853622.2 | |
| AF548543.1 | 11 | 1081 | AAQ12343.1 | |
| AK006819.1 | 11 | 1080 | ||
| BC052356.1 | 11 | 1089 | AAH52356.1 |

Genomic Map