Assay Details
Target Gene Details
Entrez Gene ID: | 277154 |
Gene Name: | NYN domain and retroviral integrase containing |
Gene Aliases: |
AA409373, C80530, mKIAA1305 |
Location: |
Chr.14:55854115-55874736 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nynrin | NM_001040072.1 | NP_001035161.1 | ||
| AK220380.1 | BAD90437.1 | |||
| BC151043.1 |
More Information
Set Membership: |
|

Genomic Map