Assay Details
Target Gene Details
Entrez Gene ID: | 67295 |
Gene Name: | RAB3C, member RAS oncogene family |
Gene Aliases: |
2700062I01Rik, 3110015B08Rik, 3110037E15Rik, AI850886 |
Location: |
Chr.13:110054187-110281041 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab3c | NM_023852.5 | NP_076341.1 | ||
| AB232589.1 | BAF02851.1 | |||
| AF312037.1 | AAG60047.1 | |||
| AJ310532.1 | CAC32042.1 | |||
| AK012468.1 | ||||
| AK014050.1 | BAB29132.1 | |||
| AK014132.1 | BAB29172.1 | |||
| AK079574.1 | BAC37689.1 | |||
| AK148904.1 | BAE28694.1 | |||
| AK161006.1 | BAE36145.1 | |||
| AK164316.1 | BAE37733.1 | |||
| AY026947.1 | AAK08980.1 | |||
| BC114335.2 | AAI14336.1 | |||
| CJ123980.1 |

Genomic Map