Assay Details
Target Gene Details
Entrez Gene ID: | 20184 |
Gene Name: | ubiquitin interaction motif containing 1 |
Gene Aliases: |
9430016E08Rik, D330018D10Rik, D630032M02Rik, RIP110, Rxrip110 |
Location: |
Chr.13:55027880-55100309 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uimc1 | NM_001293660.1 | NP_001280589.1 | ||
| NM_011307.2 | NP_035437.1 | |||
| NR_121592.1 | ||||
| AK052704.1 | BAC35106.1 | |||
| AK078206.1 | BAC37172.1 | |||
| AK083342.1 | BAC38875.1 | |||
| AK160926.1 | BAE36094.1 | |||
| AK161177.1 | ||||
| BC037092.1 | AAH37092.1 | |||
| BC040808.1 | AAH40808.1 |

Genomic Map