Assay Details
Target Gene Details
Entrez Gene ID: | 218103 |
Gene Name: | solute carrier family 17 (sodium phosphate), member 2 |
Gene Aliases: |
C730032N17Rik, NPT3 |
Location: |
Chr.13:23806941-23825191 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc17a2 | NM_144836.2 | NP_659085.1 | ||
| AK040717.1 | ||||
| AK050184.1 | ||||
| AK050277.1 | ||||
| BC018306.1 | AAH18306.1 |
More Information
Set Membership: |
|

Genomic Map