Assay Details
Target Gene Details
Entrez Gene ID: | 621976 |
Gene Name: | transmembrane protein 170B |
Gene Aliases: |
EG621976 |
Location: |
Chr.13:41606216-41641357 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem170b | NM_001163572.1 | NP_001157044.1 |
More Information
Set Membership: |
|

Genomic Map