Assay Details
Target Gene Details
Entrez Gene ID: | 66392 |
Gene Name: | prolactin family 2, subfamily b, member 1 |
Gene Aliases: |
2310047B08Rik, PLP-K, Plpk, Prlpk, mPLP-K |
Location: |
Chr.13:27383345-27390846 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prl2b1 | NM_025532.3 | 2 | 195 | NP_079808.1 |
| AF525155.1 | 2 | 146 | AAN39703.1 | |
| AK005411.1 | 2 | 196 | BAB24009.1 | |
| AK009863.1 | 2 | 196 | ||
| BC100329.1 | 2 | 185 | AAI00330.1 | |
| BC111112.1 | 2 | 191 | AAI11113.1 | |
| BC138199.1 | 2 | 188 |
More Information
Set Membership: |
|

Genomic Map