Assay Details
Target Gene Details
Entrez Gene ID: | 19356 |
Gene Name: | RAD17 homolog (S. pombe) |
Gene Aliases: |
MmRad24 |
Location: |
Chr.13:100617164-100651061 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad17 | NM_001044371.2 | NP_001037836.1 | ||
| NM_001283011.1 | NP_001269940.1 | |||
| NM_011233.3 | NP_035363.2 | |||
| AF085737.1 | AAC36335.1 | |||
| AJ011923.1 | CAA09868.1 | |||
| AK134277.1 | BAE22079.1 | |||
| AK162016.1 | ||||
| AK167358.1 | ||||
| BC066855.1 | AAH66855.1 |

Genomic Map