Assay Details
Target Gene Details
Entrez Gene ID: | 66475 |
Gene Name: | ribosomal protein S23 |
Gene Aliases: |
2410044J15Rik |
Location: |
Chr.13:90923122-90924732 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps23 | NM_024175.3 | NP_077137.1 | ||
| AK166994.1 | BAE39173.1 | |||
| BY043509.1 |
More Information
Set Membership: |
|

Genomic Map