Assay Details
Target Gene Details
Entrez Gene ID: | 105355 |
Gene Name: | solute carrier family 17 (sodium phosphate), member 3 |
Gene Aliases: |
AW261723, Npt4 |
Location: |
Chr.13:23836918-23860718 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc17a3 | NM_001164743.1 | NP_001158215.1 | ||
| NM_134069.3 | NP_598830.2 | |||
| AK128896.1 | ||||
| AK143896.1 | ||||
| BC018252.1 | AAH18252.1 | |||
| BC024459.1 | AAH24459.1 | |||
| BC034179.1 | AAH34179.1 | |||
| BC034183.1 | AAH34183.1 |
More Information
Set Membership: |
|

Genomic Map