Assay Details
Target Gene Details
Entrez Gene ID: | 107448 |
Gene Name: | unc-5 homolog A (C. elegans) |
Gene Aliases: |
Unc5h1, mKIAA1976 |
Location: |
Chr.13:54949417-55006018 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unc5a | NM_153131.3 | NP_694771.1 | ||
| AJ487852.1 | CAD32250.1 | |||
| AK122575.1 | 7 | 2322 | BAC65857.1 | |
| BC058084.1 | AAH58084.1 |
More Information
Set Membership: |
|

Genomic Map