Assay Details
Target Gene Details
Entrez Gene ID: | 67881 |
Gene Name: | magnesium-dependent phosphatase 1 |
Gene Aliases: |
1810034K20Rik, AI035604 |
Location: |
Chr.14:55657879-55660508 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mdp1 | NM_023397.4 | NP_075886.1 | ||
| NR_028316.1 | ||||
| AF230273.1 | AAK00763.1 | |||
| AK007319.1 | BAB24954.1 | |||
| AK007694.1 | BAB25194.1 | |||
| AK008881.1 | BAB25949.1 | |||
| AK160438.1 | BAE35788.1 | |||
| BC046613.1 | AAH46613.1 | |||
| CJ176407.1 |
More Information
Set Membership: |
|

Genomic Map