Assay Details
Target Gene Details
Entrez Gene ID: | 69134 |
Gene Name: | family with sequence similarity 25, member C |
Gene Aliases: |
2200001I15Rik, Fam25 |
Location: |
Chr.14:34351882-34355393 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam25c | NM_183278.2 | NP_899101.1 | ||
| AK008614.1 | BAC25225.1 | |||
| BC147104.1 | ||||
| BC147105.1 |

Genomic Map