Assay Details
Target Gene Details
Entrez Gene ID: | 26363 |
Gene Name: | biotinidase |
Gene Aliases: |
- |
Location: |
Chr.14:31640999-31668197 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btd | NM_025295.4 | NP_079571.1 | ||
| AK005506.1 | BAB24086.1 | |||
| AK135375.1 | BAE22509.1 | |||
| BC024051.1 |
More Information
Set Membership: |
|

Genomic Map