Assay Details
Target Gene Details
Entrez Gene ID: | 56044 |
Gene Name: | v-ral simian leukemia viral oncogene homolog A (ras related) |
Gene Aliases: |
3010001O15Rik, AW322615, Ral, Rasl1 |
Location: |
Chr.13:17880575-17944217 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rala | NM_019491.5 | NP_062364.3 | ||
| AF244951.1 | AAG23136.1 | |||
| AK013881.1 | BAB29033.1 | |||
| BC031741.1 | AAH31741.1 | |||
| Z48587.1 | CAA88488.1 |
More Information
Set Membership: |
|

Genomic Map