Assay Details
Target Gene Details
Entrez Gene ID: | 56705 |
Gene Name: | RAN binding protein 9 |
Gene Aliases: |
IBAP-1, Ibap1, RanBPM |
Location: |
Chr.13:43402673-43481283 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ranbp9 | NM_019930.2 | NP_064314.2 | ||
| AF006465.1 | AAD01272.1 | |||
| AK132714.1 | BAE21317.1 |
More Information
Set Membership: |
|

Genomic Map