Assay Details
Target Gene Details
Entrez Gene ID: | 268656 |
Gene Name: | serine palmitoyltransferase, long chain base subunit 1 |
Gene Aliases: |
AW552086, C77762, E030036H05, Lcb1 |
Location: |
Chr.13:53332748-53377417 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptlc1 | NM_009269.2 | NP_033295.2 | ||
| AF003823.1 | AAC02264.1 | |||
| AK028713.1 | BAE20444.1 | |||
| AK053207.1 | BAC35310.1 | |||
| AK079578.1 | BAC37690.1 | |||
| AK084391.1 | BAC39172.1 | |||
| AK084445.1 | BAC39185.1 | |||
| AK089030.1 | BAC40709.1 | |||
| AK161140.1 | BAE36210.1 | |||
| BC046323.1 | AAH46323.1 | |||
| X95641.1 | CAA64897.1 |
More Information
Set Membership: |
|

Genomic Map