Assay Details
Target Gene Details
Entrez Gene ID: | 67000 |
Gene Name: | prolactin family 3, subfamily a, member 1 |
Gene Aliases: |
1600016E11Rik, PLP-H, PLP-I, Plpi, Prlpi |
Location: |
Chr.13:27259489-27276660 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prl3a1 | NM_025896.2 | 2 | 316 | NP_080172.1 |
| AF525154.1 | 2 | 301 | AAN39702.1 | |
| AK005474.1 | 2 | 317 | BAB24064.1 | |
| AK005567.1 | 2 | 314 | ||
| BC100334.1 | 2 | 253 | AAI00335.1 |
More Information
Set Membership: |
|

Genomic Map