Assay Details
Target Gene Details
Entrez Gene ID: | 27372 |
Gene Name: | prolactin family 3, subfamily c, member 1 |
Gene Aliases: |
PLP-J, Prlpi, Prlpj, decidualin |
Location: |
Chr.13:27196660-27203749 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prl3c1 | NM_001163218.1 | NP_001156690.1 | ||
| NM_013766.2 | NP_038794.1 | |||
| AB019118.1 | BAA85988.1 | |||
| AF150740.1 | AAF13035.1 | |||
| AF234637.1 | AAF40436.1 | |||
| AK014980.1 | ||||
| BC120576.1 | ||||
| BC120578.1 |
More Information
Set Membership: |
|

Genomic Map