Assay Details
Target Gene Details
Entrez Gene ID: | 76789 |
Gene Name: | mitotic spindle organizing protein 1 |
Gene Aliases: |
2410129H14Rik, AI120158, Mozart1 |
Location: |
Chr.14:99034544-99046463 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mzt1 | NM_175245.4 | NP_780454.1 | ||
| AK010788.1 | ||||
| AK047481.1 | 1 | 84 | ||
| AK082817.1 | BAC38633.1 | |||
| AK087564.1 | ||||
| AK150268.1 | BAE29424.1 | |||
| AK151232.1 | BAE30224.1 | |||
| AK151358.1 | BAE30333.1 | |||
| AK151819.1 | BAE30716.1 | |||
| AK160911.1 | BAE36085.1 | |||
| AK165605.1 | BAE38289.1 | |||
| AK168114.1 | BAE40085.1 | |||
| AK169036.1 | BAE40827.1 | |||
| AK170746.1 | BAE41999.1 | |||
| BB844588.1 | ||||
| BC117005.1 | ||||
| BC117007.1 |

Genomic Map