Assay Details
Target Gene Details
Entrez Gene ID: | 110902 |
Gene Name: | cholinergic receptor, nicotinic, alpha polypeptide 2 (neuronal) |
Gene Aliases: |
Acra-2, Acra2, BC011490 |
Location: |
Chr.14:66133238-66153396 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrna2 | NM_144803.2 | NP_659052.1 | ||
| AY574261.1 | AAS90357.1 | |||
| BC011490.1 | AAH11490.1 |
More Information
Set Membership: |
|

Genomic Map