Assay Details
Target Gene Details
Entrez Gene ID: | 219072 |
Gene Name: | HAUS augmin-like complex, subunit 4 |
Gene Aliases: |
9430093H08Rik, AU016300, D14Ertd500e |
Location: |
Chr.14:54541785-54554361 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Haus4 | NM_145462.2 | NP_663437.2 | ||
| AK079161.1 | BAC37564.1 | |||
| AK079852.1 | BAC37767.1 | |||
| AK087969.1 | ||||
| BC004644.1 | AAH04644.1 | |||
| BC011095.1 | AAH11095.1 |

Genomic Map