Assay Details
Target Gene Details
Entrez Gene ID: | 105504 |
Gene Name: | exocyst complex component 5 |
Gene Aliases: |
AI447711, AI448003, Gm76, PRO1912, SEC10, Sec10l1 |
Location: |
Chr.14:49011922-49066667 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exoc5 | NM_207214.3 | NP_997097.1 | ||
| AK134562.1 | ||||
| AK152307.1 | BAE31113.1 | |||
| AK157473.1 | BAE34093.1 | |||
| AK164067.1 | BAE37611.1 | |||
| AK167145.1 | BAE39290.1 | |||
| AK170148.1 | ||||
| AK171012.1 | BAE42183.1 | |||
| BC049967.1 | AAH49967.1 |
More Information
Set Membership: |
|

Genomic Map