Assay Details
Target Gene Details
Entrez Gene ID: | 286940 |
Gene Name: | filamin, beta |
Gene Aliases: |
AL024016, Fln-b |
Location: |
Chr.14:7817957-7951588 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Flnb | NM_001081427.1 | 8 | 1479 | NP_001074896.1 |
| NM_134080.1 | 8 | 1479 | NP_598841.1 | |
| BP753071.1 | 2 | 296 | ||
| CD555839.1 | 2 | 269 |

Genomic Map