Assay Details
Target Gene Details
Entrez Gene ID: | 219114 |
Gene Name: | spindle and kinetochore associated complex subunit 3 |
Gene Aliases: |
F630043A04Rik |
Location: |
Chr.14:57806560-57826190 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ska3 | NM_198605.3 | NP_941007.1 | ||
| AK089223.1 | BAC40798.1 | |||
| AK138255.1 | 1 | 1252 | ||
| AK166171.1 | ||||
| AK199129.1 | ||||
| AK217625.1 | ||||
| BC117500.1 | ||||
| BC117501.1 |

Genomic Map