Assay Details
Target Gene Details
Entrez Gene ID: | 16426 |
Gene Name: | inter-alpha trypsin inhibitor, heavy chain 3 |
Gene Aliases: |
AW108094, Intin3, Itih-3 |
Location: |
Chr.14:30908572-30923755 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itih3 | NM_008407.2 | NP_032433.2 | ||
| AK146700.1 | ||||
| BC015276.1 | AAH15276.1 | |||
| X70393.1 | CAA49843.1 |
More Information
Set Membership: |
|

Genomic Map