Assay Details
Target Gene Details
Entrez Gene ID: | 619301 |
Gene Name: | transmembrane protein 253 |
Gene Aliases: |
G630016D24Rik |
Location: |
Chr.14:52015275-52019787 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem253 | NM_001033805.3 | NP_001028977.1 | ||
| AK144313.1 | BAE25829.1 | |||
| BC147689.1 |
Target Gene Details
Entrez Gene ID: | 69890 |
Gene Name: | zinc finger protein 219 |
Gene Aliases: |
2010302A17Rik |
Location: |
Chr.14:52006075-52020733 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp219 | NM_001253694.1 | NP_001240623.1 | ||
| NM_001253696.1 | NP_001240625.1 | |||
| NM_027248.2 | NP_081524.2 | |||
| AB063578.1 | BAB61057.1 | |||
| AK154332.1 | ||||
| BC096486.1 | AAH96486.1 |
More Information
Set Membership: |
|

Genomic Map