Assay Details
Target Gene Details
Entrez Gene ID: | 27389 |
Gene Name: | dual specificity phosphatase 13 |
Gene Aliases: |
DUSP13A, DUSP13B, Gm1203, LMW-DSP6, MDSP, TMDP, TS-DSP6 |
Location: |
Chr.14:21733395-21748622 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dusp13 | NM_001007268.1 | 3 | 493 | NP_001007269.1 |
| AY674052.1 | 3 | 493 | AAT79357.1 |

Genomic Map