Assay Details
Target Gene Details
Entrez Gene ID: | 27389 |
Gene Name: | dual specificity phosphatase 13 |
Gene Aliases: |
DUSP13A, DUSP13B, Gm1203, LMW-DSP6, MDSP, TMDP, TS-DSP6 |
Location: |
Chr.14:21733395-21748622 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dusp13 | NM_001007268.1 | 6 | 981 | NP_001007269.1 |
| NM_013849.3 | 2 | 306 | NP_038877.2 | |
| AB027003.1 | 1 | 87 | BAA89411.1 | |
| AF237620.1 | 2 | 289 | AAK15037.1 | |
| AK006247.1 | 2 | 307 | BAB24480.2 | |
| AY674052.1 | 6 | 981 | AAT79357.1 | |
| BC100421.1 | 1 | 212 | AAI00422.1 |

Genomic Map