Assay Details
Target Gene Details
Entrez Gene ID: | 105428 |
Gene Name: | family with sequence similarity 149, member B |
Gene Aliases: |
AA536717, AI428773, Fam149b1, mKIAA0974 |
Location: |
Chr.14:20348162-20383491 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam149b | NM_001024512.2 | 12 | 1719 | NP_001019683.1 |
| NM_172379.3 | 10 | 1440 | NP_759011.2 | |
| AK129252.1 | 9 | 1119 | BAC98062.1 | |
| BC008267.1 | 6 | 783 | ||
| BC057907.1 | 10 | 1342 | AAH57907.1 | |
| BC069846.1 | 12 | 1617 | AAH69846.1 | |
| BC080849.1 | 12 | 1659 | AAH80849.1 |

Genomic Map