Assay Details
Target Gene Details
Entrez Gene ID: | 105428 |
Gene Name: | family with sequence similarity 149, member B |
Gene Aliases: |
AA536717, AI428773, Fam149b1, mKIAA0974 |
Location: |
Chr.14:20348162-20383491 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam149b | NM_001024512.2 | NP_001019683.1 | ||
| NM_001177633.1 | NP_001171104.1 | |||
| NM_172379.3 | NP_759011.2 | |||
| AK031973.1 | BAC27633.1 | |||
| AK037808.1 | ||||
| AK077495.1 | ||||
| AK129252.1 | BAC98062.1 | |||
| BB622169.1 | ||||
| BC025183.1 | AAH25183.1 | |||
| BC057907.1 | AAH57907.1 | |||
| BC069846.1 | AAH69846.1 | |||
| BC080849.1 | AAH80849.1 |

Genomic Map