Assay Details
Target Gene Details
Entrez Gene ID: | 219148 |
Gene Name: | family with sequence similarity 167, member A |
Gene Aliases: |
A030013D21 |
Location: |
Chr.14:63436394-63465502 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam167a | NM_177628.4 | 2 | 288 | NP_808296.2 |
| BB840230.1 | 2 | 288 | ||
| BC065085.1 | AAH65085.1 |

Genomic Map