Assay Details
Target Gene Details
Entrez Gene ID: | 13829 |
Gene Name: | dematin actin binding protein |
Gene Aliases: |
AI325486, Epb4.9, Epb49, dematin |
Location: |
Chr.14:70602184-70636048 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmtn | NM_001252662.1 | 9 | 1153 | NP_001239591.1 |
| NM_001252663.1 | 9 | 1153 | NP_001239592.1 | |
| NM_001252664.1 | 8 | 980 | NP_001239593.1 | |
| NM_001252665.1 | 8 | 879 | NP_001239594.1 | |
| NM_001252666.1 | 7 | 912 | NP_001239595.1 | |
| NM_013514.4 | 10 | 1094 | NP_038542.1 | |
| AF079846.3 | 8 | 823 | AAD34233.1 | |
| AF155547.1 | 8 | 823 | AAD38412.1 | |
| AK143840.1 | 9 | 1147 | ||
| AK149301.1 | 8 | 790 | BAE28799.1 | |
| AK155927.1 | 9 | 1148 | ||
| AK158744.1 | 8 | 911 | BAE34638.1 | |
| AK162866.1 | 8 | 831 | BAE37092.1 | |
| AK165294.1 | 9 | 1145 | BAE38123.1 | |
| BC016897.1 | 10 | 1094 | AAH16897.1 | |
| BC037021.1 | 7 | 899 | AAH37021.1 |
More Information
Set Membership: |
|

Genomic Map