Assay Details
Target Gene Details
Entrez Gene ID: | 218952 |
Gene Name: | fermitin family homolog 2 (Drosophila) |
Gene Aliases: |
AA960555, Kindlin-2, Mig2, Plekhc1 |
Location: |
Chr.14:45458792-45530113 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fermt2 | NM_146054.2 | NP_666166.2 | ||
| AK145030.1 | BAE26194.1 | |||
| AK165608.1 | ||||
| AK166552.1 | BAE38850.1 | |||
| BC033436.1 | AAH33436.1 | |||
| BC034168.1 | AAH34168.1 | |||
| CD550909.1 |

Genomic Map