Assay Details
Target Gene Details
Entrez Gene ID: | 14171 |
Gene Name: | fibroblast growth factor 17 |
Gene Aliases: |
- |
Location: |
Chr.14:70636203-70643014 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgf17 | NM_008004.4 | 3 | 631 | NP_032030.1 |
| AB009250.1 | 3 | 135 | BAA25430.1 | |
| AK077555.1 | 3 | 633 | BAC36859.1 | |
| AK082858.1 | 3 | 522 | BAC38656.1 | |
| BC119033.1 | 3 | 617 | ||
| BC144776.1 |
More Information
Set Membership: |
|

Genomic Map