Assay Details
Target Gene Details
Entrez Gene ID: | 56794 |
Gene Name: | 2-hydroxyacyl-CoA lyase 1 |
Gene Aliases: |
1600020H07Rik, Hpcl, Phyh2 |
Location: |
Chr.14:31607226-31641286 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hacl1 | NM_019975.3 | 12 | 1148 | NP_064359.2 |
| AJ132139.1 | 12 | 1146 | CAB65550.1 | |
| AK005505.1 | 12 | 1135 | BAB24085.1 | |
| AK041686.1 | 12 | 1144 | BAC31032.1 | |
| AK050078.1 | 12 | 1148 | BAC34059.1 | |
| BC021360.1 | 12 | 1079 | AAH21360.1 |
More Information
Set Membership: |
|

Genomic Map