Assay Details
Target Gene Details
Entrez Gene ID: | 108671 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 9 |
Gene Aliases: |
5330419I01Rik, AU020082 |
Location: |
Chr.14:20384638-20388910 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc9 | NM_134081.5 | NP_598842.1 | ||
| AK075979.1 | BAC36092.1 | |||
| AK077324.1 | BAC36750.1 | |||
| AK153033.1 | BAE31665.1 | |||
| AK162075.1 | BAE36712.1 | |||
| BC014686.1 | AAH14686.2 | |||
| BC023787.1 | AAH23787.1 | |||
| BC027012.1 | AAH27012.1 |
More Information
Set Membership: |
|

Genomic Map