Assay Details
Target Gene Details
Entrez Gene ID: | 50527 |
Gene Name: | ERO1-like (S. cerevisiae) |
Gene Aliases: |
ERO1-L |
Location: |
Chr.14:45283087-45318572 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ero1l | NM_015774.3 | NP_056589.1 | ||
| AK009667.1 | BAB26428.1 | |||
| AK142595.1 | BAE25121.1 | |||
| BC025102.1 | AAH25102.1 | |||
| CT010195.1 | CAJ18403.1 | |||
| CT010259.1 | CAJ18467.1 |
More Information
Set Membership: |
|

Genomic Map