Assay Details
Target Gene Details
Entrez Gene ID: | 16391 |
Gene Name: | interferon regulatory factor 9 |
Gene Aliases: |
Irf-9, Isgf3g, p48 |
Location: |
Chr.14:55603985-55610030 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Irf9 | NM_001159417.1 | 4 | 664 | NP_001152889.1 |
| NM_001159418.1 | 3 | 462 | NP_001152890.1 | |
| NM_008394.3 | 3 | 578 | NP_032420.1 | |
| AK043697.1 | 1 | 39 | ||
| AK145911.1 | 2 | 330 | BAE26744.1 | |
| AK149742.1 | 2 | 329 | ||
| BB860586.1 | ||||
| BC005435.1 | 4 | 439 | AAH05435.1 | |
| BC012968.1 | 3 | 473 | AAH12968.1 | |
| CK788642.1 | 3 | 405 | ||
| U51992.1 | 3 | 382 | AAC52494.1 |
More Information
Set Membership: |
|

Genomic Map