Assay Details
Target Gene Details
Entrez Gene ID: | 16391 |
Gene Name: | interferon regulatory factor 9 |
Gene Aliases: |
Irf-9, Isgf3g, p48 |
Location: |
Chr.14:55603985-55610030 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Irf9 | NM_001159417.1 | NP_001152889.1 | ||
| NM_001159418.1 | NP_001152890.1 | |||
| NM_008394.3 | NP_032420.1 | |||
| AK043697.1 | 3 | 490 | ||
| AK145911.1 | BAE26744.1 | |||
| AK149742.1 | ||||
| BC005435.1 | AAH05435.1 | |||
| BC012968.1 | AAH12968.1 | |||
| CK788642.1 | ||||
| U51992.1 | AAC52494.1 |
More Information
Set Membership: |
|

Genomic Map