Assay Details
Target Gene Details
Entrez Gene ID: | 219132 |
Gene Name: | PHD finger protein 11D |
Gene Aliases: |
D14Ertd668e, Phf11, Phf11l |
Location: |
Chr.14:59347407-59370520 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phf11d | NM_199015.4 | NP_950180.3 | ||
| AK156583.1 | BAE33765.1 | |||
| BC030186.1 | AAH30186.1 | |||
| BC145673.1 | ||||
| EU155107.1 |
More Information
Set Membership: |
|

Genomic Map