Assay Details
Target Gene Details
Entrez Gene ID: | 286940 |
Gene Name: | filamin, beta |
Gene Aliases: |
AL024016, Fln-b |
Location: |
Chr.14:7817957-7951588 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Flnb | NM_001081427.1 | 24 | 4294 | NP_001074896.1 |
| NM_134080.1 | 24 | 4294 | NP_598841.1 | |
| BU703688.1 | 4 | 527 | ||
| CN534523.1 | 2 | 71 |

Genomic Map