Assay Details
Target Gene Details
Entrez Gene ID: | 791403 |
Gene Name: | myosin heavy chain associated RNA transcript |
Gene Aliases: |
D830015G02Rik, ENSMUSG00000053118, Myheart |
Location: |
Chr.14:54968787-54974349 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mhrt | NR_033497.1 | |||
| AK052874.1 |
Target Gene Details
Entrez Gene ID: | 140781 |
Gene Name: | myosin, heavy polypeptide 7, cardiac muscle, beta |
Gene Aliases: |
B-MHC, MyHC-I, Myhc-b, Myhcb, beta-MHC |
Location: |
Chr.14:54970688-54994549 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 39 - Exon 40 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myh7 | NM_080728.2 | NP_542766.1 | ||
| AK147031.1 | BAE27623.1 | |||
| AK168593.1 | ||||
| AK168677.1 | BAE40526.1 | |||
| AK169198.1 | BAE40973.1 | |||
| AK169371.1 | BAE41119.1 | |||
| AY056464.1 | AAL17913.1 | |||
| BC121789.1 | ||||
| BC158070.1 | ||||
| M74752.1 |

Genomic Map