Assay Details
Target Gene Details
Entrez Gene ID: | 268822 |
Gene Name: | aarF domain containing kinase 5 |
Gene Aliases: |
A230108P17 |
Location: |
Chr.15:76576359-76595812 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adck5 | NM_172960.3 | NP_766548.2 | ||
| NR_073123.1 | ||||
| AK039217.1 | BAC30280.1 | |||
| AK163557.1 | ||||
| BC060528.1 | AAH60528.1 | |||
| BQ886383.1 |
More Information
Set Membership: |
|

Genomic Map